Human Pancreatic Organoids for Cancer Research Applications
Read more about
- What is Pancreatic Ductal Adenocarcinoma?
- Pancreatic Ductal Adenocarcinoma Organoid Biobank
- Pancreatic Ductal Adenocarcinoma Organoid Data
What is Pancreatic Ductal Adenocarcinoma?
Pancreatic ductal adenocarcinoma (PDAC) represents 85% of all cases of pancreatic cancer. Pancreatic cancer is the seventh most common cancer with an incidence rate of approximately 3.5% and is the fourth most common cause of cancer-related death for both sexes and all ages. Smoking, diabetes mellitus, obesity, age, and genetic predisposition are the main risk factors identified so far to be implicated in the development of PDAC.
The KRAS mutations G12D and G12V are found in more than 90% of PDAC cases. The success of drugs targeting KRASG12C suggests the potential for drugs specifically targeting these alternative PDAC-associated KRAS mutations.
Historically, pancreatic cancer cell lines or animal models have been used to study PDAC but are limited by lack of biological relevance or high costs. Patient derived organoids (PDOs) are novel in vitro 3D cell models that have tissue-like functionality and are an alternative to traditional 2D cell lines and PDX animal models.
Pancreatic Ductal Adenocarcinoma Organoid Biobank
The 3dGRO® pancreatic organoid biobank is a collection of tissue-derived human pancreatic cancer organoids (PDAC organoids) for disease modeling research. The pancreatic organoid biobank contains highly characterized organoid lines from PDAC diseased patients, representing primary tumors containing wild-type and KRAS driver mutations.
For complete pancreatic organoid culture protocols and media recipes visit the 3dGRO Human Pancreatic Organoid User Guide.
Catalog Number | ID | Disease | Source | Age | Sex | KRAS Mutation |
---|---|---|---|---|---|---|
SCC700 | PPTO.02 | Pancreatic Ductal Adenocarcinoma | Primary Tumor | 62 | M | G12D |
SCC702 | PPTO.13 | Pancreatic Ductal Adenocarcinoma | Primary Tumor | 73 | M | G12R |
SCC703 | PPTO.26 | Pancreatic Ductal Adenocarcinoma | Primary Tumor | 81 | M | G12V |
SCC704 | PPTO.30 | Pancreatic Ductal Adenocarcinoma | Primary Tumor | 73 | M | G12D |
SCC705 | PPTO.36 | Pancreatic Ductal Adenocarcinoma | Primary Tumor | 76 | F | G12D |
SCC706 | PPTO.46 | Pancreatic Ductal Adenocarcinoma | Primary Tumor | 66 | F | G12R |
SCC707 | PPTO.49 | Pancreatic Ductal Adenocarcinoma | Primary Tumor | 55 | M | G12V |
SCC708 | PPTO.61 | Pancreatic Ductal Adenocarcinoma | Primary Tumor | 60 | F | Wild-Type |
SCC709 | PPTO.64 | Pancreatic Ductal Adenocarcinoma | Primary Tumor | 58 | M | G12D |
SCC710 | PPTO.65 | Pancreatic Ductal Adenocarcinoma | Primary Tumor | 69 | F | G12D |
SCC711 | PPTO.76 | Pancreatic Ductal Adenocarcinoma | Primary Tumor | 57 | F | G12D |
SCC712 | PPTO.80 | Pancreatic Ductal Adenocarcinoma | Primary Tumor | 78 | F | G12R |
SCC713 | PPTO.90 | Pancreatic Ductal Adenocarcinoma | Primary Tumor | 57 | F | G12V |
SCC714 | PPTO.93 | Pancreatic Ductal Adenocarcinoma | Primary Tumor | 66 | M | Q61H |
SCC716 | OCIP.174 | Pancreatic Ductal Adenocarcinoma | Xenograph | 59 | F | Q61H |
SCC717 | OCIP.180 | Pancreatic Ductal Adenocarcinoma | Xenograph | 64 | M | Wild-Type |
SCC718 | OCIP.255 | Pancreatic Ductal Adenocarcinoma | Xenograph | 66 | F | Unknown |
SCC719 | OCIP.285 | Pancreatic Ductal Adenocarcinoma | Xenograph | 65 | F | Q61L |
SCC720 | OCIP.335 | Pancreatic Ductal Adenocarcinoma | Xenograph | 74 | M | Wild-Type |
Figure 1.Time course of PDAC cancer organoid growth over a 5-day period shows positive cell proliferation after thaw (SCC700, PPTO.02) at 2X (top images) or 10X (bottom images) magnification.
Figure 2.Human PDAC cancer organoid (SCC706, PPTO.46) morphology and confluency prior to cryopreservation on day 6 (first image at 2X magnification, second image at 10X magnification).
Figure 3.Immunocytochemical characterization of human pancreatic organoids (SCC700, PPTO.02). Human PDAC cancer organoids are positive for Cytokeratin 19 (CK19, green), epithelial cell adhesion molecule (EpCAM, green) and Actin (red). Cells counterstained with DAPI (blue).
Network error: Failed to fetch
3dGRO™ organoids were derived utilizing HUB Organoid Technology.