Primary Human Intrahepatic Biliary Epithelial Cells (Cholangiocytes) Culture Protocol
What are Primary Human Cholangiocytes?
Primary human Cholangiocytes, also known as human intrahepatic biliary epithelial cells, are epithelial cells that line the intrahepatic bile ducts. Cholangiocytes are important for ductal bile modification and are targeted in multiple liver diseases, such as primary biliary cirrhosis, cholangiocarcinoma and sclerosing cholangitis.
Our frozen human Cholangiocytes cells are derived from the human liver, with each donor providing documented consent for research use of non-transplantable organs or tissues. Following the primary culture, these cells are cryopreserved. Each lot undergoes testing for specific cell markers and comes with a guarantee of ≥ 70% post-thaw viability. This protocol provides a step-by-step demonstration of how to thaw, plate, and cultivate primary human Cholangiocytes.
Primary Human Cholangiocytes Culture Materials
- Normal Human Intrahepatic Biliary Epithelial Cells (Cholangiocytes)
Note: Upon receipt, immediately store cryovial(s) in vapor phase liquid nitrogen. - Collagen Type I, Rat Tail
- Tissue culture treated multiwell plates
- Dulbecco’s PBS, without Calcium & Magnesium (DPBS)
- Trypsin
Preparing Primary Human Cholangiocyte Medium
Formulations for human hepatic stellate cell media are readily available from literature. Below is an example media from one of the publications.1
Components | Catalog Number | Working Stock | Final Dilution | Final Conc. | Final Volume (mL) |
---|---|---|---|---|---|
DMEM/F-12 mix |
|
| 1x | 439 | |
FBS |
|
| 10% | 50 | |
Hydrocortisone | 1mg/mL** | 500x | 2µg/mL | 1 | |
Insulin | 10mg/mL | 2,326x | 4.3µg/mL | 0.215 | |
Cholera Toxin | 0.1mg/mL | 10,000x | 10ng/mL | 0.05 | |
Triiodo-L-thyronine | 20ug/mL (30uM)*** | 15,000x | 2nM | 0.033 | |
Human EGF | 0.1mg/mL | 10,000 | 10ng/mL | 0.05 | |
L-glutamine | 200mM | 100x | 2mM, 1x | 5 | |
Pen/Strep | 100x | 100x | 1x | 5 | |
Total volume | 500 |
**Dissolve in 100% EtOH
***To prepare 20 µg/ml stock solution: add 1ml 1N NaOH per mg 3,3', 5-triiodo-L-thyronine; gently swirl to dissolve. To this, add 49 ml sterile medium per ml 1N NaOH added.
Human Cholangiocyte Culture Method
Unless otherwise specified, this protocol was performed in a Class II laminar flow biohood with an aspirator. Incubators are set to 37°C and 5% CO2 and researchers should wear PPE such as lab coats, gloves, and safety glasses.
Thawing and Plating Human Cholangiocytes
- Place vial in a 37°C water bath, holding and rotating the vial gently until the contents are completely thawed. Remove the vial from the water bath immediately, wipe dry, rinse the vial with 70% ethanol and transfer to a sterile field. Remove the cap, being careful not to touch the interior threads with fingers.
- Using a pipette, gently transfer contents of vial to a 15 ml conical tube containing 5 mL of Cholangiocyte medium. Wash vial with 1 mL of the medium and add the wash to the same conical tube.
- Centrifuge tube at 300xg for 5 minutes. After centrifugation, aspirate medium and re-suspend the contents in medium.
- For expansion, seed the cells at a density of 5,000 cells/cm2 on collagen I coated plates.
- For best results, do not disturb the culture for at least 12 hours after seeding. Change the medium the next day to remove any residual DMSO or unattached cells.
- Feed cells fresh Cholangiocyte medium every other day until ready for assay or expansion.
Sub-Culturing Human Cholangiocytes
- Subculture cells when they have reached 70 - 80% confluency. Cells can be monitored with cell imaging instruments such as the Millicell® DCI Digital Cell Imager.
- Warm Cholangiocytes medium in a 37˚C water bath.
Note: Make sure the 0.25% trypsin solution and the Dulbecco’s Phosphate Buffered Saline without Calcium & Magnesium (DPBS) are at room temperature. - Aspirate the medium and rinse cells with DPBS. Add trypsin solution into flask and incubate in a 37˚C incubator for 3-5 minutes or until the cells detach.
- As soon as the cells detach, wash cells from flask using 2X the volume with Cholangiocyte medium. Transfer to centrifuge tube and centrifuge at 250xg for 5 minutes. After centrifugation aspirate the medium, re-suspend the cells, and count for seeding.
- Seed the cells at a density of 5,000 cells/cm2 in collagen I coated plates.
Network error: Failed to fetch