Merck
  • Discovery and mechanism of action of a novel series of apoptosis inducers with potential vascular targeting activity.
Discovery and mechanism of action of a novel series of apoptosis inducers with potential vascular targeting activity.
Molecular cancer therapeutics (2004-11-16)
Shailaja Kasibhatla, Henriette Gourdeau, Karen Meerovitch, John Drewe, Sanjeeva Reddy, Ling Qiu, Hong Zhang, Frederick Bergeron, David Bouffard, Quan Yang, John Herich, Serge Lamothe, Sui Xiong Cai, Ben Tseng
ZUSAMMENFASSUNG
A novel series of 2-amino-4-(3-bromo-4,5-dimethoxy-phenyl)-3-cyano-4H-chromenes was identified as apoptosis-inducing agents through our cell-based apoptosis screening assay. Several analogues from this series, MX-58151, MX-58276, MX-76747, MX-116214, MX-126303, and MX-116407, were synthesized and further characterized. MX-116407, a lead compound from this series, induced apoptosis with an EC50 of 50 nmol/L and inhibited cell growth with a GI50 of 37 nmol/L in T47D breast cancer cells. Treatment of cells with these analogues led to G2-M arrest, cleavage of essential proapoptotic caspase substrates, and induction of nuclear fragmentation. We identified these compounds as tubulin destabilizers with binding site at or close to the colchicine binding site. Compounds in this series were also active in drug-resistant cancer cell lines with a GI50 value for one of the analogues (MX-58151) of 2.5 nmol/L in paclitaxel-resistant, multidrug-resistant MES-SA/DX5 tumor cells. This series of compounds displayed high selectivity against proliferating versus resting cells. Interestingly, these compounds were shown to disrupt preformed endothelial cell capillary tubules in vitro and affect functional vasculature to induce tumor necrosis in vivo and are thus likely to work as tumor vasculature targeting agents. Among these compounds, MX-116407 showed capillary tubule disruption activity in vitro at concentrations well below the cytotoxic dose. In a separate study, we further characterized the antitumor efficacy and pharmacokinetic profile of this series of compounds and identified MX-116407 as a potent apoptosis-inducing agent with apparent activity as tumor vasculature targeting agent.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Discovery® Cyano HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® Cyano Supelguard Kartusche, 5 μm particle size, L × I.D. 2 cm × 4 mm