Merck
21978-U
Swagelok® Verbinder an NPT-Innengewinde
Swagelok® 200-7-4, brass, 1/8 in. Swagelok, 1/4 in. female NPT
Synonym(e):
Swagelok® 200-7-4, MESSING ADAPTER, 1/8TX1/4P, PK2
About This Item
UNSPSC-Code:
41112500
NACRES:
SB.54
Materialien
brass
Verpackung
pkg of 2 ea
Hersteller/Markenname
Swagelok® 200-7-4
Fitting
1/4 in. female NPT
1/8 in. Swagelok
Allgemeine Beschreibung
Mit Nuts und Ferrules.
Rechtliche Hinweise
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Analysenzertifikate (COA)