Merck
Z708259
Foam plugs
size 46-50 mm
Synonym(e):
foam plug
About This Item
UNSPSC-Code:
41100000
Größe
46-50 mm
Allgemeine Beschreibung
Reusable, nonabsorbent plugs provide a snug fit yet allow free airflow. Easily manipulate to fit flasks or tubes. Plugs spring back to original shape after repeated use. Autoclavable.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)