Merck
Z380164
Sigma-Aldrich
Security bag for X-ray film
For 14 × 17 inch (35 × 43 cm) film
About This Item
UNSPSC-Code:
24111500
Allgemeine Beschreibung
Contains and protects a whole box of x-ray film in a light-tight environment. The fabric is tightly woven black nylon coated inside with opaque black vinyl. The overlapping flap closure secures the bag on all edges with a hook-and-loop seal.