Merck
Z363030
Sigma-Aldrich
Kodak® BioMax® MS film
Synonym(e):
autoradiography film, xray film
About This Item
UNSPSC-Code:
41105341
NACRES:
NB.22
Größe
7 in. (18 cm) × 9.5 in. (24 cm)
Allgemeine Beschreibung
Double-emulsion film specifically engineered for biotechnology applications. Provides maximum sensitivity for detection of 32P and 125I in Northern and Southern blots, reducing quantity of isotope needed.
Sheets of film with no interleaved paper
Rechtliche Hinweise
BioMax is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)