TMO3664-0500
Nalgene® economy graduated cylinder
polypropylene, volume 500 mL, case of 8 ea
Synonym(e):
PP graduated cylinder, PP graduated flask
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
polypropylene
Verpackung
case of 8 ea
Hersteller/Markenname
Nalgene 3664-0500
Kapazität
500 mL
Volumen
500 mL
Verwandte Kategorien
Allgemeine Beschreibung
Dispense liquids easily with simple to read, molded-in graduations that won′t rub off. Affordably priced Thermo Scientific Nalgene Polypropylene Economy Graduated Cylinders have a generous pour spout and exhibit excellent chemical and break resistance.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)