Merck
EPM1282-9915
Platform for Eppendorf® New Brunswick Innova® 44/44R Incubator Shakers
for 1 L Erlenmeyer flasks, universal, non-Interchangeable, aluminum (heavy duty)
About This Item
UNSPSC-Code:
41103816
NACRES:
NB.73
Materialien
aluminum (heavy duty)
Leistungsmerkmale
for 1 L Erlenmeyer flasks
non-Interchangeable
universal
Hersteller/Markenname
Eppendorf® M1282-9915 (Global)
Verwandte Kategorien
Allgemeine Beschreibung
Platforms to fit the Eppendorf New Brunswick 44 and 44 R incubated shakers.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)