Merck
EP951020541
Eppendorf® twin.tec PCR Plates 384
Optimized rim for barcodes, PCR clean, yellow, pack of 300 ea
About This Item
UNSPSC-Code:
41106300
Materialien
conical bottom
polypropylene wells
yellow
Sterilität
non-sterile
Leistungsmerkmale
PCR clean
lid: no
skirt
Verpackung
pack of 300 ea
Hersteller/Markenname
Eppendorf® 951020541
Größe
384 well
Well-Volumen
45 μL
Eignung
suitable for PCR
Verwandte Kategorien
Allgemeine Beschreibung
  • Eppendorf® twin.tec PCR Plate 384, 45 µL, PCR clean, colorless, 300 plates
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler pro 384
  • 40 µL max. well volume
  • Available with barcode (upon request), for skirted and semi-skirted only
Eppendorf® twin.tec PCR Plate 384, 45 µL, PCR clean, yellow, 300 plates
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler pro 384
  • 40 µL max. well volume
Leistungsmerkmale und Vorteile
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler pro 384
  • 40 µL max. well volume
  • Available with barcode (upon request), for skirted and semi-skirted only
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)