Merck
EP022639911
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
FA-45-48-11, aerosol-tight
About This Item
UNSPSC-Code:
41103909
Materialien
aluminum
Leistungsmerkmale
RCF 19,083 × g
aerosol-tight
Verpackung
pack of 1 ea
Hersteller/Markenname
Eppendorf® 022639911
Kompatibilität
5804/5804R & 5810/5810R
FA-45-48-11
Verwandte Kategorien
Allgemeine Beschreibung
Rotor FA-45-48-11, aerosol-tight
  • 48 places for 1.5/2.0 mL tubes
  • Max. speed: 19,083 × g (max. 13,000 rpm)1)
  • Eppendorf QuickLock® rotor lid for fast lid locking
  • Aerosol-tight for safe centrifugation of hazardous samples2)
  • Anodized aluminum for high chemical resistance
  • Rotor and lid are autoclavable
1) Please note that a software upgrade may be necessary for older centrifuges when using this rotor. Please contact your local Eppendorf organization or dealer for further information. 2) Aerosol-tightness tested by Public Health England, Porton Down, Great Britain.
Leistungsmerkmale und Vorteile
48 places for 1.5/2.0 mL tubes
Max. speed: 19,083 × g (max. 13,000 rpm)1)
Eppendorf QuickLock® rotor lid for fast lid locking
Aerosol-tight for safe centrifugation of hazardous samples2)
Anodized aluminum for high chemical resistance
Rotor and lid are autoclavable
1) Please note that a software upgrade may be necessary for older centrifuges when using this rotor. Please contact your local Eppendorf organization or dealer for further information. 2) Aerosol-tightness tested by Public Health England, Porton Down, Great Britain.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf QuickLock is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)