Merck
CLS6770-RTA25
Corning® LSE Adapter
0.25 for rotor (Cat. No. 6770-RT)
Synonym(e):
Corning® LSE Adapters
About This Item
UNSPSC-Code:
23152104
NACRES:
SB.56
Leistungsmerkmale
0.25 for rotor (Cat. No. 6770-RT)
Hersteller/Markenname
Corning 6770-RTA.25
Eignung
suitable for (for use with Corning LSE compact centrifuge)
Allgemeine Beschreibung
These are optional adapters for Corning LSE compact centrifuge rotors: CLS480136, CLS480137, CLS480138; CLS480139
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.