Merck
AXYVT38410ULRS
Pipette tips in 384-tip format, robotic
size 10 μL, pkg of 5x3840tips/cs, racked, sterile
Synonym(e):
pipet tips
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.02
Sterilität
sterile
Hersteller/Markenname
Axygen Inc VT-384-10UL-R-S
Corning
Verpackung
pkg of 5x3840tips/cs
Größe
10 μL
Allgemeine Beschreibung
  • Axygen® automation pipette tips in 384-tip format are designed for Agilent®/Velocity11 VPrep® and Bravo 96LT, 8LT, 384ST, 96ST, and 16ST head.
  • Manufactured to stringent specifications including automated assembly to minimize contamination risks and ensure consistent performance and quality.
  • The clear polypropylene high-quality tips are ideal for sensitive biological applications.
  • RNase-/DNase-free and nonpyrogenic.
Sonstige Hinweise
Browse all pipette tips
Rechtliche Hinweise
Agilent is a registered trademark of Agilent Technologies, Inc.
Axygen is a registered trademark of Corning, Inc.
Velocity11 VPrep is a registered trademark of Corning, Inc.