Merck
AXYPCR96FSBK
PCR microplate, full skirt
size 96 wells, conical bottom black polypropylene wells, pkg of 5x10plates/cs
Synonym(e):
96 multiwell plate
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
Materialien
conical bottom black polypropylene wells
Sterilität
non-sterile
Hersteller/Markenname
Axygen Inc PCR-96-FS-BK
Corning
Verpackung
pkg of 5x10plates/cs
Maximales Volumen
200 μL
Größe
96 wells
Arbeitsvolumen
200 μL
Bindungsart
non-treated surface