Merck
AXYPCR48B
Corning® Axygen® PCR Microplate
size 48 wells, conical bottom blue polypropylene wells, pkg of 5x10plates/cs
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
Materialien
conical bottom blue polypropylene wells
Sterilität
non-sterile
Hersteller/Markenname
Axygen Inc PCR-48-B
Corning
Verpackung
pkg of 5x10plates/cs
Maximales Volumen
200 μL
Größe
48 wells
Arbeitsvolumen
200 μL
Eignung
suitable for PCR
Bindungsart
non-treated surface
Rechtliche Hinweise
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)