Merck
AXYPCR38455Y
Corning® Axygen® PCR Microplate
size 384 wells, conical bottom yellow polypropylene wells, maximum volume 55 μL, pkg of 5x10plates/cs
Synonym(e):
384 multiwell plate
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
Materialien
conical bottom yellow polypropylene wells
Sterilität
non-sterile
Hersteller/Markenname
Axygen Inc PCR-384-55-Y
Corning
Verpackung
pkg of 5x10plates/cs
Maximales Volumen
55 μL
Größe
384 wells
Arbeitsvolumen
50 μL
Eignung
suitable for (thermal cycling)
Bindungsart
non-treated surface
Rechtliche Hinweise
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)