Z685097
Ace sparger with hose barb
L × O.D. 915 mm × 14 mm, porosity 70-100 μm
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Hersteller/Markenname
Ace Glass 6452-44
L × AD
915 mm × 14 mm
Absperrhahngröße
0-5 mm
Porengröße
70-100 μm porosity
Allgemeine Beschreibung
Sparger tube with Ace fritted bottoms are available in various porosities. Top has either 0-3 or 0-5 mm stopcock and a 90 degree side arm hose connection. 10 mm O.D. spargers fit inside #11 Ace-Threds, 14 mm O.D. within #15 Ace- Threds.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)