Merck
Z654809
IKA® VORTEX 3 attachments
VG 3.31 test tube attachment (for VG 3.3), for 54 Eppendorf tubes, 1/cs
About This Item
UNSPSC-Code:
41103814
NACRES:
NB.73
Verpackung
case of 1
pack of 1
Hersteller/Markenname
IKA 3344300
Allgemeine Beschreibung
Note: Use with IKA VORTEX 3 attachment VG 3.3 (Z644795)
Rechtliche Hinweise
IKA is a registered trademark of IKA-Werke GmbH & Co. KG