Merck
Z555770
Ace reaction flask head
capacity 2,000-4,000 mL, center joint: ST/NS 45/50, side Joints: ST/NS 24/40 (2)
Synonym(e):
flask lab, lab flask, reaction flasks
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
glass
three-neck (in-line)
Verbindung
center joint: ST/NS45/50
side Joints: ST/NS24/40 (2)
Sterilität
non-sterile
Verpackung
pack of 1
Hersteller/Markenname
Ace Glass 651228
Kapazität
2,000-4,000 mL