Merck
Z419958
Sigma-Aldrich
Aldrich® addition funnels with PTFE needle valve
Jacketed, capacity 250 mL, joint: ST/NS 24/40
Synonym(e):
funnel glass, glass funnel, glass funnels
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.43
Verbindung
joint: ST/NS24/40
Höhe
500 mm
Kapazität
250 mL
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC