Merck
Z401846
MSA respirator goggles
About This Item
UNSPSC-Code:
46182002
Allgemeine Beschreibung
Clear, soft frame, 4-vent goggles are specially constructed to wear with MSA respirators. Goggles have a 0.060in. thick, clear, PC antifog lens. Elastic headband.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)