Z255866
Latex-Laborschlauch
I.D. × O.D. 5/16 in. × 7/16 in.
Alle Fotos(1)
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.35
Beschreibung
standard
Verpackung
pkg of 100 feet
ID × AD
5/16 in. × 7/16 in.
Wandabmessungen
1/16 in.
Allgemeine Beschreibung
High-purity, seamless, amber tubing for general-purpose use with liquids and gases. Durometer: 35 ±5. Talc-free.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)