Merck
Z158291
Ace Enlarging Adapter
female joint: SJ 35/20, male joint: ST/NS 24/40
Synonym(e):
glass adapter, glass adapters
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
Verbindung
female joint: SJ35/20
male joint: ST/NS24/40
Hersteller/Markenname
Ace Glass 502025