WHA10308210
Whatman® filter paper, Grade 989
circles, diam. 110 mm, pack of 100
Synonym(e):
Z749028
Alle Fotos(1)
About This Item
UNSPSC-Code:
41104900
NACRES:
NB.24
Materialien
circles
Verpackung
pack of 100
Hersteller/Markenname
Whatman 10308210
Whatman Article No. 28414152 (US reference)
Parameter
medium flow rate
Durchmesser
110 mm
Porengröße
8-10 μm
Sonstige Hinweise
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)