TEM-FCF200NIUA
Ultrathin Formvar/Carbon Supported Nickel Square Mesh
size 200 mesh, box of 50
Synonym(e):
FORMVAR/CARBON SQUARE MESH - NI 200 MESH UA 50/BX
Alle Fotos(1)
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Empfohlene Produkte
Verpackung
box of 50
Größe
200 mesh
Allgemeine Beschreibung
Ultrathin 5-6nm Formvar and 1nm Carbon Supported Square Mesh Grids: Nickel Grid, 200 mesh. A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)