Merck
SYNS312044
Sigma-Aldrich
Synthware T-bore glass stopcock Entire assembly
bore size 6 mm, plug size 20/44
Synonym(e):
Synthware S312044, t-shape stopcock
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware S312044
Bohrungsgröße
6 mm
Verwandte Kategorien
Allgemeine Beschreibung
With solid glass plug. With constant pressure retention spring and clip.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.