Merck
SYNA871922
Synthware 75 degree angled drying tube
joint: ST/NS 19/22
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Materialien
colorless tube
glass tube
Verbindung
joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A871922
Allgemeine Beschreibung
Angled at 75° to avoid desiccant contact with reactions.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)