Merck
SYNA234250
Synthware anti-splash adapter with fritted disc
250 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 24/40
Synonym(e):
Synthware A234250, rotary evaporator bump trap
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
bottom joint: ST/NS24/40
joint: ST/NS top24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A234250
Kapazität
250 mL
Allgemeine Beschreibung
With extra coarse fritted disc sealed above lower inner joint. Frit reduces foam from being carryover.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.