SYNA112440
Synthware™ modified Claisen adapter
joint: ST/NS 24/40
Synonym(e):
Synthware A112440, distilling adapter
Empfohlene Produkte
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A112440
Allgemeine Beschreibung
Three-inlet type for multiple entries into a single neck flask. One side joint is at a 45° angle.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.