Merck
SYNA111420
Synthware modified Claisen adapter
joint: ST/NS 14/20
Synonym(e):
Synthware A111420, distilling adapter
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A111420
Allgemeine Beschreibung
Three-inlet type for multiple entries into a single neck flask. One side joint is at a 45° angle.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.