GF87544604
Copper-beryllium alloy, Cu98Be2
foil, 4mm disks, 0.125mm thickness, half hard
Synonym(e):
CU070350
Alle Fotos(1)
About This Item
UNSPSC-Code:
31152304
NACRES:
NA.23
Form
foil (disks - half hard)
Hersteller/Markenname
Goodfellow 875-446-04
Größe × Dicke
4 mm × 0.125 mm
Allgemeine Beschreibung
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)