Merck
GF55922632
Phosphor bronze alloy, Cu94Sn6
sheet, 150 x 150mm, 3.2mm thickness, hard
Synonym(e):
CU053150
About This Item
UNSPSC-Code:
11101711
NACRES:
NA.23
Form
foil
Hersteller/Markenname
Goodfellow 559-226-32
Größe × Dicke
150x150 mm × 3.2 mm