Merck
253103
Sigma-Aldrich
29H,31H-Phthalocyanin
β-form, 98%
About This Item
Empirische Formel (Hill-System):
C32H18N8
CAS-Nummer:
Molekulargewicht:
514.54
Beilstein:
378323
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12352103
PubChem Substanz-ID:
NACRES:
NA.23
Qualitätsniveau
Assay
98%
Form
powder
mp (Schmelzpunkt)
>300 °C (dec.) (lit.)
λmax
698 nm
SMILES String
c1ccc2c3nc(nc4[nH]c(nc5nc(nc6[nH]c(n3)c7ccccc67)c8ccccc58)c9ccccc49)c2c1
InChI
1S/C32H18N8/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25/h1-16H,(H2,33,34,35,36,37,38,39,40)
InChIKey
IEQIEDJGQAUEQZ-UHFFFAOYSA-N
Anwendung
Suitable as laser dye
Lagerklassenschlüssel
11 - Combustible Solids
WGK
WGK 1
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Persönliche Schutzausrüstung
Eyeshields, Gloves, type N95 (US)
Die Dokumentenbibliothek aufrufen
Yasuhiro Maruoka et al.
Vaccines, 8(3) (2020-09-18)
Near infrared photoimmunotherapy (NIR-PIT) is a newly developed and highly selective cancer treatment that induces necrotic/immunogenic cell death. It employs a monoclonal antibody (mAb) conjugated to a photo-absorber dye, IRDye700DX, which is activated by NIR light. Tumor-targeting NIR-PIT is also
Teofil Jesionowski et al.
Marine drugs, 16(3) (2018-03-10)
The biosynthesis, chemistry, structural features and functionality of spongin as a halogenated scleroprotein of keratosan demosponges are still paradigms. This review has the principal goal of providing thorough and comprehensive coverage of spongin as a naturally prefabricated 3D biomaterial with
David Rand et al.
Advanced materials (Deerfield Beach, Fla.), 30(25), e1707292-e1707292 (2018-05-03)
An efficient nanoscale semiconducting optoelectronic system is reported, which is optimized for neuronal stimulation: the organic electrolytic photocapacitor. The devices comprise a thin (80 nm) trilayer of metal and p-n semiconducting organic nanocrystals. When illuminated in physiological solution, these metal-semiconductor
Lukasz Bodek et al.
Beilstein journal of nanotechnology, 11, 821-828 (2020-06-20)
The adsorption behavior of tin phthalocyanine (SnPc) molecules on rutile TiO2(110) was studied by scanning tunneling microscopy (STM). Low-temperature STM measurements of single molecules reveal the coexistence of two conformations of molecules on the TiO2 surface. Density functional theory-based simulations
Qinggong Tang et al.
Journal of controlled release : official journal of the Controlled Release Society, 260, 154-163 (2017-06-12)
Photoimmunotherapy (PIT) is an emerging low side effect cancer therapy based on a monoclonal antibody (mAb) conjugated with a near-infrared (NIR) phthalocyanine dye IRDye 700DX. IR700 is fluorescent, can be used as an imaging agent, and also is phototoxic. It
Artikel
Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.