Merck
  • Effects of intranasally dosed posaconazole on fungal load and biomarkers in Aspergillus fumigatus infected immunocompromised mice.
Effects of intranasally dosed posaconazole on fungal load and biomarkers in Aspergillus fumigatus infected immunocompromised mice.
Mycoses (2017-07-13)
Genki Kimura, Takahiro Nakaoki, Yuki Nishimoto, Yuto Suzuki, Garth Rapeport, Pete Strong, Kazuhiro Ito, Yasuo Kizawa
ABSTRACT
Although anti-fungal triazoles are dosed orally or systemically for Aspergillus fumigatus infection, systemic adverse events and limited exposure of the lung cavity would make a topical treatment for the lung an attractive option. In this study, we examined the effects of intranasally dosed posaconazole on survival rates and biomarkers in A. fumigatus (itraconazole susceptible: ATCC13073 [Af]; or resistant: NCPF7100 [AfR]) infected, temporarily neutropenic A/J mice. Once daily treatment produced a dose-dependent improvement of survival of Af-infected mice (ED
Product Description
Supelco
Posaconazole, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Silver Stain (Modified GMS) Kit
Pricing and availability is not currently available.