Merck
  • Activation of the PI3K/Akt/mTOR and MAPK Signaling Pathways in Response to Acute Solar-Simulated Light Exposure of Human Skin.
Activation of the PI3K/Akt/mTOR and MAPK Signaling Pathways in Response to Acute Solar-Simulated Light Exposure of Human Skin.
Cancer prevention research (Philadelphia, Pa.) (2015-06-03)
Yira Bermudez, Steven P Stratton, Clara Curiel-Lewandrowski, James Warneke, Chengcheng Hu, George T Bowden, Sally E Dickinson, Zigang Dong, Ann M Bode, Kathylynn Saboda, Christine A Brooks, Emanuel F Petricoin, Craig A Hurst, David S Alberts, Janine G Einspahr
ABSTRACT
The incidence of skin cancer is higher than all other cancers and continues to increase, with an average annual cost over $8 billion in the United States. As a result, identifying molecular pathway alterations that occur with UV exposure to strategize more effective preventive and therapeutic approaches is essential. To that end, we evaluated phosphorylation of proteins within the PI3K/Akt and MAPK pathways by immunohistochemistry in sun-protected skin after acute doses of physiologically relevant solar-simulated ultraviolet light (SSL) in 24 volunteers. Biopsies were performed at baseline, 5 minutes, 1, 5, and 24 hours after SSL irradiation. Within the PI3K/Akt pathway, we found activation of Akt (serine 473) to be significantly increased at 5 hours while mTOR (serine 2448) was strongly activated early and was sustained over 24 hours after SSL. Downstream, we observed a marked and sustained increase in phospho-S6 (serine 235/S236), whereas phospho-4E-BP1 (threonines 37/46) was increased only at 24 hours. Within the MAPK pathway, SSL-induced expression of phospho-p38 (threonine 180/tyrosine 182) peaked at 1 to 5 hours. ERK 1/2 was observed to be immediate and sustained after SSL irradiation. Phosphorylation of histone H3 (serine 10), a core structural protein of the nucleosome, peaked at 5 hours after SSL irradiation. The expression of both p53 and COX-2 was increased at 5 hours and was maximal at 24 hours after SSL irradiation. Apoptosis was significantly increased at 24 hours as expected and indicative of a sunburn-type response to SSL. Understanding the timing of key protein expression changes in response to SSL will aid in development of mechanistic-based approaches for the prevention and control of skin cancers.
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
SAFC
L-Threonine
Sigma-Aldrich
Glycine ethyl ester hydrochloride, 99%
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Anti-p53 (Ab-6) (Pantropic) Mouse mAb (DO-1), liquid, clone DO-1, Calbiochem®
Sigma-Aldrich
Anti-PCNA (Ab-1) Mouse mAb (PC10), liquid, clone PC10, Calbiochem®
Sigma-Aldrich
Phosphatase substrate, Suitable for manufacturing of diagnostic kits and reagents
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Hematoxylin