Merck
  • Characterization of sophorolipid biosynthetic enzymes from Starmerella bombicola.
Characterization of sophorolipid biosynthetic enzymes from Starmerella bombicola.
FEMS yeast research (2015-08-25)
Karen M J Saerens, Inge N A Van Bogaert, Wim Soetaert
ABSTRACT
Altering glycolipid structure by genetic engineering of Starmerella bombicola is a recently started research topic and worthy alternative to the unsuccessful selective feeding strategies conventionally applied to reach this goal. One question to be addressed when expressing heterologous proteins in S. bombicola is the activity of the subsequent biosynthetic enzymes toward such modified substrates. In this scope, we studied the substrate specificity of the UDP-glucosyltransferases UgtA1 and UgtB1, responsible for the stepwise synthesis of sophorolipids from a hydroxylated fatty acid, and that of the acetyltransferase, responsible for acetylation of the sophorolipid molecule. All enzymes showed specificity toward a C18:1 chained acceptor and both glucosyltransferases were highly selective toward the UDP-glucose donor. Severe product inhibition of the glucosyltransferases explains the limited accumulation of sophorolipid intermediates by earlier created single deletion mutants of S. bombicola. Finally, a more detailed study of the acetylation of sophorolipid intermediates sheds light on the enzymatic cascade during synthesis.
Sigma-Aldrich
Adenosine-5′-diphosphoglucose disodium salt, ≥93%
Sigma-Aldrich
Guanosine 5′-diphospho-D-mannose sodium salt from Saccharomyces cerevisiae, Type I, ≥97% (HPLC)
Sigma-Aldrich
Uridine 5′-diphosphogalactose disodium salt, ≥97.0%
Sigma-Aldrich
12-Hydroxydodecanoic acid, 97%
Sigma-Aldrich
16-Hydroxyhexadecanoic acid, 98%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)