Merck
  • Enzymatic incorporation of an azide-modified UTP analog into oligoribonucleotides for post-transcriptional chemical functionalization.
Enzymatic incorporation of an azide-modified UTP analog into oligoribonucleotides for post-transcriptional chemical functionalization.
Nature protocols (2012-05-12)
Harita Rao, Arun A Tanpure, Anupam A Sawant, Seergazhi G Srivatsan
ABSTRACT
This protocol describes the detailed experimental procedure for the synthesis of an azide-modified uridine triphosphate analog and its effective incorporation into an oligoribonucleotide by in vitro transcription reactions. Furthermore, procedures for labeling azide-modified oligoribonucleotides post-transcriptionally with biophysical probes by copper(I)-catalyzed alkyne-azide cycloaddition (CuAAC) and Staudinger reactions are also provided. This post-transcriptional chemical modification protocol is simple and modular, and it affords labeled oligonucleotides in reasonable amounts for biophysical assays. The procedure for enzymatic incorporation of the monophosphate of azide-modified UTP into an oligoribonucleotide transcript takes ∼2 d, and subsequent post-transcriptional chemical functionalization of the transcript takes about 2 d.
Sigma-Aldrich
Bromophenol Blue sodium salt, for molecular biology, suitable for electrophoresis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Tris(3-hydroxypropyltriazolylmethyl)amine, 95%
Sigma-Aldrich
Sodium methoxide, reagent grade, 95%, powder
Sigma-Aldrich
Thioacetamide, reagent grade, 98%