Merck
  • [Specifics of hormonal and energy balance in patients with hyperplasia and endometrial neoplasia with metabolic syndrome in the background].
[Specifics of hormonal and energy balance in patients with hyperplasia and endometrial neoplasia with metabolic syndrome in the background].
Voprosy onkologii (2013-07-03)
A L Chernyshova, L A Kolomiets, N V Bochkarëva, I V Kondakova
ABSTRACT
We conducted a comparative investigation of the hormonal status (LH, FSH, estradiol, progesterone, testosterone, prolactin, SHBG), energy status (leptin, ghrelin, insulin), and carbohydrate and lipid metabolism in patients with endometrial hyperplasia and neoplasia (168 patients) with or without metabolic syndrome in the background. Patients with metabolic syndrome had a high frequency of elevated estrogen (72%), testosterone (65%), insulin (81%), leptin (68%). There was a marked increase in the basal level of luteinizing hormone, prolactin, index, LH/FSH, but decrease in FSH and progesterone. There were significant changes in carbohydrate and lipid metabolism. The possible mechanisms for the contribution of the investigated factors to the development of the pathological processes in the endometrium are presented.
Product Description
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Progesterone, meets USP testing specifications
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
Follicle Stimulating Hormone from human pituitary, ~7,000 IU/mg (powder)
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Testosterone, European Pharmacopoeia (EP) Reference Standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Prolactin human, recombinant, expressed in E. coli, lyophilized powder, BioReagent, suitable for cell culture, >97% (SDS-PAGE)
Sigma-Aldrich
Estradiol, meets USP testing specifications
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Progesterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®