Merck
  • A new approach for diffusive sampling based on SPME for occupational exposure assessment.
A new approach for diffusive sampling based on SPME for occupational exposure assessment.
Journal of occupational and environmental hygiene (2013-01-30)
P Marín, J F Periago, C Prado
ABSTRACT
Passive sampling is a well-established methodology for the evaluation of exposures to environmental volatile organic compounds (VOCs). The solid-phase microextraction (SPME) technique is a reliable means of sampling VOCs in air. SPME is also being applied as a passive sampler to determine time-weighted average exposure. The use of SPME as a diffusive sampler was evaluated. The passive sampler is based on the use of a cylindrical diffusion cell (porous hydrophobic polyethylene) with an 80 μm carboxen/polydimethylsiloxane fiber to obtain radial diffusion of the analytes to the sorbent. Standard atmospheres of organic vapors in air were used to determine the experimental uptake rates for toluene and chlorobenzene. Toluene concentrations between 2 and 38 mg/m(3) with sampling times between 15 and 60 min were evaluated, as well as chlorobenzene concentrations between 2 and 47 mg/m(3) with sampling times between 30 and 60 min. The mean diffusive uptake rate was 2.14 mL/min for toluene and 2.57 mL/min for chlorobenzene, and no statistical significant effects of concentration and sampling time were observed under the studied conditions for the two compounds. Relative standard deviation ranged from 2.6 to 6.5%. The performance of the sampler under varying concentrations of toluene was also tested, showing that the sampler reflects the average exposure concentration. Effects of temperature, relative humidity, velocity of the air, back diffusion, competitive adsorption, and the stability of chlorobenzene in the sampler were also studied. Sampler behavior was tested in gas stations, and the results were successfully compared with a 3M-3500 diffusive sampler. The results are promising for using this new SPME device for diffusive monitoring for occupational exposure assessment.
Product Description
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4 in. × 3 1/2 in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea
Supelco
Carboxen® 569, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Toluene, analytical standard
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
ORBO 78 HBr on Carboxen® 564 specially cleaned (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Chlorobenzene, analytical standard
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Supelco
Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Supelco
SPME Fiber Assembly, matrix active group 50/30µm DVB/CAR/PDMS, StableFlex fiber, manual holder, needle size 24 ga
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME Fiber Assembly, for use with autosampler, matrix active group 50/30µm DVB/CAR/PDMS, needle size 23 ga, metal alloy fiber, fiber L 2 cm