Merck
  • Highly efficient capture and long-term encapsulation of dye by catanionic surfactant vesicles.
Highly efficient capture and long-term encapsulation of dye by catanionic surfactant vesicles.
Langmuir : the ACS journal of surfaces and colloids (2006-07-13)
Xiang Wang, Emily J Danoff, Nikolai A Sinkov, Jae-Ho Lee, Srinivasa R Raghavan, Douglas S English
ABSTRACT
Vesicles formed from the cationic surfactant, cetyltrimethylammonium tosylate (CTAT) and the anionic surfactant, sodium dodecylbenzenesulfonate (SDBS), were used to sequester the anionic dye carboxyfluorescein. Carboxyfluorescein was efficiently sequestered in CTAT-rich vesicles via two mechanisms: encapsulation in the inner water pool and electrostatic adsorption to the charged bilayer. The apparent encapsulation efficiency (22%) includes both encapsulated and adsorbed fractions. Entrapment of carboxyfluorescein by SDBS-rich vesicles was not observed. Results show the permeability of the catanionic membrane is an order of magnitude lower than that of phosphatidylcholine vesicles and the loading capacity is more than 10 times greater.
Sigma-Aldrich
Hexadecyltrimethylammonium p-toluenesulfonate