Merck
  • Thyroid Hormone Receptor Beta Inhibits PI3K-Akt-mTOR Signaling Axis in Anaplastic Thyroid Cancer via Genomic Mechanisms.
Thyroid Hormone Receptor Beta Inhibits PI3K-Akt-mTOR Signaling Axis in Anaplastic Thyroid Cancer via Genomic Mechanisms.
Journal of the Endocrine Society (2021-07-15)
Cole D Davidson, Eric L Bolf, Noelle E Gillis, Lauren M Cozzens, Jennifer A Tomczak, Frances E Carr
ABSTRACT
Thyroid cancer is the most common endocrine malignancy, and the global incidence has increased rapidly over the past few decades. Anaplastic thyroid cancer (ATC) is highly aggressive, dedifferentiated, and patients have a median survival of fewer than 6 months. Oncogenic alterations in ATC include aberrant phosphoinositide 3 kinase (PI3K) signaling through receptor tyrosine kinase (RTK) amplification, loss of phosphoinositide phosphatase expression and function, and protein kinase B (Akt) amplification. Furthermore, the loss of expression of the tumor suppressor thyroid hormone receptor beta (TRβ) is strongly associated with ATC. TRβ is known to suppress PI3K in follicular thyroid cancer and breast cancer by binding to the PI3K regulatory subunit p85α. However, the role of TRβ in suppressing PI3K signaling in ATC is not completely delineated. Here we report that TRβ indeed suppresses PI3K signaling in ATC cell lines through unreported genomic mechanisms, including a decrease in RTK expression and an increase in phosphoinositide and Akt phosphatase expression. Furthermore, the reintroduction and activation of TRβ in ATC cell lines enables an increase in the efficacy of the competitive PI3K inhibitors LY294002 and buparlisib on cell viability, migration, and suppression of PI3K signaling. These findings not only uncover additional tumor suppressor mechanisms of TRβ but shed light on the implication of TRβ status and activation on inhibitor efficacy in ATC tumors.
Sigma-Aldrich
Anti-Thyroid Hormone Receptor β-1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-PI3 Kinase Antibody, p85, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-phospho-Glycogen Synthase(Ser641/Ser645) Antibody, Upstate®, from rabbit