Merck
  • Fusarium verticillioides: evaluation of fumonisin production and effect of fungicides on in vitro inhibition of mycelial growth.
Fusarium verticillioides: evaluation of fumonisin production and effect of fungicides on in vitro inhibition of mycelial growth.
Mycopathologia (2010-06-29)
Virgínia Carla A Falcão, Mario Augusto Ono, Tatiana de Ávila Miguel, Edio Vizoni, Elisa Yoko Hirooka, Elisabete Yurie Sataque Ono
ABSTRACT
The objectives of this study were to evaluate the fumonisin production by 16 F. verticillioides strains on corn cultures and the effect of quintozene and fludioxonil + metalaxyl-M fungicides on "in vitro" mycelial growth on agar. In addition, the effect of fludioxonil + metalaxyl-M on fumonisin production in defined liquid culture medium was analyzed. Fumonisin B(1) levels on corn cultures ranged from 2.41 to 3996.36 μg/g and the F. verticillioides 103F strain produced the highest level (3996.36 ± 390.49 μg/g, P < 0.05). F. verticillioides strains were inoculated in potato dextrose agar with the addition of quintozene (75 to 9,375 μg/ml) and fludioxonil + metalaxyl-M (1.5 + 0.6 to 187.5 + 75 μg/ml) in order to evaluate the effect of these fungicides on "in vitro" mycelial growth. The F. verticillioides strains showed great variability concerning ED(50) values, which were below the recommended application dose for quintozene, but above that for fludioxonil + metalaxyl-M. Moreover, fungicide addition to the culture medium increased mean FB(1) levels compared to the control, suggesting the importance of focusing on the effect of fungicides on mycotoxin production as well as on the phytopathogen control.
Product Description
Supelco
Quintozene, PESTANAL®, analytical standard
Supelco
Pentachloronitrobenzene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Fludioxonil, PESTANAL®, analytical standard
Sigma-Aldrich
Pentachloronitrobenzene, ≥94%