![Phaclofen solid](/deepweb/assets/sigmaaldrich/product/structures/338/212/072d974c-74b0-4c7e-b1ec-92616fd0122c/640/072d974c-74b0-4c7e-b1ec-92616fd0122c.png)
Phaclofen
![Phaclofen solid](/deepweb/assets/sigmaaldrich/product/structures/338/212/072d974c-74b0-4c7e-b1ec-92616fd0122c/640/072d974c-74b0-4c7e-b1ec-92616fd0122c.png)
Synonym(s):
3-Amino-2-(4-chlorophenyl)propanephosphonic acid
Empirical Formula (Hill Notation):
C9H13ClNO3P
CAS Number:
Molecular Weight:
249.63
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing