Nilutamide
Synonym(s):
5,5-Dimethyl-3-[4-nitro-3-(trifluoromethyl)phenyl]-2,4-imidazolidinedione, Anandron, RU-23908, Nilutamide
Empirical Formula (Hill Notation):
C12H10F3N3O4
CAS Number:
Molecular Weight:
317.22
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade