1-Deoxymannojirimycin hydrochloride
Synonym(s):
1,5-Dideoxy-1,5-imino-D-mannitol hydrochloride
Empirical Formula (Hill Notation):
C6H13NO4 · HCl
CAS Number:
Molecular Weight:
199.63
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Activity Spectrum
Mode of Action
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing