Skip to Content
Merck
All Photos(1)

Key Documents

V900800

Sigma-Aldrich

Ethylenediamine

Vetec, reagent grade, ≥98%

Synonym(s):

1,2-Diaminoethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2NH2
CAS Number:
Molecular Weight:
60.10
Beilstein:
605263
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

reagent grade

vapor density

2.07 (vs air)

vapor pressure

10 mmHg ( 20 °C)

product line

Vetec

Assay

≥98%

form

liquid

autoignition temp.

716 °F

expl. lim.

16 %

refractive index

n20/D 1.4565 (lit.)

pH

12.2 (20 °C, 110 g/L)

bp

118 °C (lit.)

mp

8.5 °C (lit.)

density

0.899 g/mL at 25 °C (lit.)

SMILES string

NCCN

InChI

1S/C2H8N2/c3-1-2-4/h1-4H2

InChI key

PIICEJLVQHRZGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1B - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Jae Shin et al.
Soft matter, 11(25), 5037-5043 (2015-05-29)
The noble vesicular system of polydiacetylene showed a red shift using two types of detecting systems. One of the systems involves the absorption of target materials from the outer side of the vesicle, and the other system involves the permeation
J Zuidema
Pharmaceutisch weekblad. Scientific edition, 7(4), 134-140 (1985-08-23)
Ethylenediamine is an excipient with many industrial and pharmaceutical uses. It is included in creams as a stabilizer and in aminophylline as the counter ion of theophylline. Ethylenediamine is one of the most frequent contact sensitizers, producing local and generalized
Leslie S Hamachi et al.
Chemical science, 10(26), 6539-6552 (2019-08-02)
We report a method to control the composition and microstructure of CdSe1-x S x
Mona Nystad et al.
American journal of obstetrics and gynecology, 211(6), 686-686 (2014-06-25)
The purpose of this study was to investigate the expression and subcellular localization of laeverin, a placenta-specific membrane-bound aminopeptidase, in preeclamptic placentas and its role in trophoblast cell migration and invasion. Expression of laeverin was investigated in 6 normal and
Yuri Choi et al.
ACS nano, 8(11), 11377-11385 (2014-10-18)
Hybrid nanoparticles composed of multiple components can offer unique opportunities for understanding the nanoscale mechanism and advanced material applications. Here, we report the synthesis of heterodimeric silver-carbon dot nanoparticles (Ag-CD NPs) where the Ag NP is grown on the surface

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service