Skip to Content
Merck
All Photos(1)

Documents

V900644

Sigma-Aldrich

1-(2-Hydroxyethyl)piperidine

Vetec, reagent grade, 98%

Synonym(s):

1-Piperidineethanol, 2-Piperidinoethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO
CAS Number:
Molecular Weight:
129.20
Beilstein:
103390
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

refractive index

n20/D 1.4804 (lit.)

bp

199-202 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

OCCN1CCCCC1

InChI

1S/C7H15NO/c9-7-6-8-4-2-1-3-5-8/h9H,1-7H2

InChI key

KZTWONRVIPPDKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongxia Cui et al.
Journal of pharmaceutical sciences, 104(10), 3395-3403 (2015-09-10)
The study was carried out to investigate the mechanism of the ion-pair strategy in modulating zaltoprofen (ZAL) skin permeability. Seven organic amines were chosen as counter ions and the formation of ion pairs was confirmed by Fourier transform infrared and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service