Skip to Content
Merck
All Photos(1)

Key Documents

V900376

Sigma-Aldrich

DL-Leucine

Vetec, reagent grade, 99%

Synonym(s):

(±)-Amino-4-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
636005
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

99%

form

powder

color

white

mp

293-296 °C (subl.) (lit.)

SMILES string

CC(C)CC(N)C(O)=O

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)

InChI key

ROHFNLRQFUQHCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masaki Kajimoto et al.
American journal of physiology. Heart and circulatory physiology, 309(7), H1157-H1165 (2015-08-02)
Nutritional energy support during extracorporeal membrane oxygenation (ECMO) should promote successful myocardial adaptation and eventual weaning from the ECMO circuit. Fatty acids (FAs) are a major myocardial energy source, and medium-chain FAs (MCFAs) are easily taken up by cell and
Farid Ichou et al.
Journal of mass spectrometry : JMS, 49(6), 498-508 (2014-06-11)
Reproducibility among different types of excitation modes is a major bottleneck in the field of tandem mass spectrometry library development in metabolomics. In this study, we specifically evaluated the influence of collision voltage and activation time parameters on tandem mass
Konstantinos A Kouremenos et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 966, 179-186 (2014-03-29)
Water supply biofilms have the potential to harbour waterborne diseases, accelerate corrosion, and contribute to the formation of tuberculation in metallic pipes. One particular species of bacteria known to be found in the water supply networks is Pseudomonas sp., with
Soura Challal et al.
ACS chemical neuroscience, 5(10), 993-1004 (2014-08-16)
Medicinal plants used for the treatment of epilepsy are potentially a valuable source of novel antiepileptic small molecules. To identify anticonvulsant secondary metabolites, we performed an in vivo, zebrafish-based screen of medicinal plants used in Southeast Asia for the treatment
Carola W N Damen et al.
Journal of lipid research, 55(8), 1772-1783 (2014-06-04)
An ultraperformance LC (UPLC) method for the separation of different lipid molecular species and lipid isomers using a stationary phase incorporating charged surface hybrid (CSH) technology is described. The resulting enhanced separation possibilities of the method are demonstrated using standards

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service