Skip to Content
Merck
All Photos(3)

Key Documents

V900300

Sigma-Aldrich

PIPES dipotassium salt

Vetec, reagent grade, 99%

Synonym(s):

1,4-Piperazinediethanesulfonic acid dipotassium salt, Piperazine-1,4-bis(2-ethanesulfonic acid) dipotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16K2N2O6S2
CAS Number:
Molecular Weight:
378.55
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

99%

form

crystalline powder

useful pH range

6.1-7.5

pKa (25 °C)

6.8

solubility

water: 0.5 g/mL, clear, colorless

SMILES string

[K+].[K+].[O-]S(=O)(=O)CCN1CCN(CC1)CCS([O-])(=O)=O

InChI

1S/C8H18N2O6S2.2K/c11-17(12,13)7-5-9-1-2-10(4-3-9)6-8-18(14,15)16;;/h1-8H2,(H,11,12,13)(H,14,15,16);;/q;2*+1/p-2

InChI key

BJQYFCAMUXGZFN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service