Skip to Content
Merck

Skip To

V800296

Petroleum ether

LR, bp 40-60 °C

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12191502
EC Number:
232-453-7
MDL number:
Grade:
LR
Bp:
30-60 °C (lit.), 40-60 °C
Vapor pressure:
25.8 psi ( 55 °C), 7.99 psi ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

LR

vapor density

2.5 (vs air)

vapor pressure

25.8 psi ( 55 °C), 7.99 psi ( 20 °C)

product line

Vetec

form

liquid

autoignition temp.

475 °F

expl. lim.

8 %

dilution

(for analytical testing)

refractive index

n20/D 1.363 (lit.)

bp

30-60 °C (lit.), 40-60 °C

density

0.64 g/mL at 25 °C (lit.)

SMILES string

[Mg+]c1ccc(cc1)C.[Br-]

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Petroleum ether


pictograms

FlameHealth hazard

signalword

Danger

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B

Storage Class

3 - Flammable liquids

flash_point_f

-56.2 °F - closed cup

flash_point_c

-49 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

8032-32-4

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters